ENA113817017 Enamine

1-(6-ethoxypyridazin-3-yl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09040534
  • Cas Number : 956728-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27169

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27169
Availability details for
Quantity
  • Availability
  • Add to Cart