ENA113817019 Enamine

ethyl 1-(6-chloropyridazin-3-yl)-5-methyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD00103043
  • Cas Number : 303144-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27171
Availability details for
Quantity
  • Availability
  • Add to Cart