ENA113817020 Enamine

3-(4-fluorophenyl)-2-(5-methyl-1H-1,2,3,4-tetrazol-1-yl)prop-2-enoic acid

  • Cas Number : 501908-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27173
Availability details for
Quantity
  • Availability
  • Add to Cart