ENA113817022 Enamine

1-(3-methoxyphenyl)-2-sulfanylideneimidazolidin-4-one

  • MDL number MFCD09347305
  • Cas Number : 929972-78-1
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27186
Availability details for
Quantity
  • Availability
  • Add to Cart