ENA113817023 Enamine

1-cyclopentyl-4-methyl-1H-pyrazolo[3,4-b]pyridin-6-yl 3-nitrobenzene-1-sulfonate

  • MDL number MFCD09040541
  • Cas Number : 929972-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27187
Availability details for
Quantity
  • Availability
  • Add to Cart