ENA113817026 Enamine

1,3,4-trimethyl-1H-pyrazolo[3,4-b]pyridin-6-yl 3-nitrobenzene-1-sulfonate

  • MDL number MFCD09040545
  • Cas Number : 929972-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27197

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27197
Availability details for
Quantity
  • Availability
  • Add to Cart