ENA113817031 Enamine

5-benzyl-3-methyl-1,6-dihydro-1,2,4-triazin-6-one

  • MDL number MFCD09040551
  • Cas Number : 15150-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27223

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27223
Availability details for
Quantity
  • Availability
  • Add to Cart