ENA113817043 Enamine

3-phenyl-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD09040531
  • Cas Number : 1022-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27155
Availability details for
Quantity
  • Availability
  • Add to Cart