ENA113817046 Enamine

3-(2,6-dimethylmorpholin-4-yl)propan-1-amine

  • MDL number MFCD08445210
  • Cas Number : 91551-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27165
Availability details for
Quantity
  • Availability
  • Add to Cart