ENA113817047 Enamine

N-(1-acetyl-2,3-dihydro-1H-indol-5-yl)-2-chloroacetamide

  • MDL number MFCD06496453
  • Cas Number : 893722-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-22964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22964
Availability details for
Quantity
  • Availability
  • Add to Cart