ENA113817052 Enamine

3-[(chloroacetyl)(4-methoxyphenyl)amino]propanamide

  • MDL number MFCD02172211
  • Cas Number : 325776-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23751

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23751
Availability details for
Quantity
  • Availability
  • Add to Cart