ENA116261335 Enamine

1-(2-chloroacetyl)-2,3-dihydro-1H-indole-5-sulfonamide

  • MDL number MFCD09040376
  • Cas Number : 929973-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23822
Availability details for
Quantity
  • Availability
  • Add to Cart