ENA116261345 Enamine

2,6-diamino-4-oxo-1,4-dihydropyrimidine-5-carboxamide

  • MDL number MFCD26143723
  • Cas Number : 103574-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27330
Availability details for
Quantity
  • Availability
  • Add to Cart