ENA116261347 Enamine

ethyl 1,3-dimethyl-6-(propan-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylate

  • MDL number MFCD09040585
  • Cas Number : 929973-69-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27338

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27338
Availability details for
Quantity
  • Availability
  • Add to Cart