ENA116261350 Enamine

N-[1-(hydrazinecarbonyl)-2-phenyleth-1-en-1-yl]acetamide

  • MDL number MFCD09040594
  • Cas Number : 15048-22-3
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27356
Availability details for
Quantity
  • Availability
  • Add to Cart