ENA116261637 Enamine

3-[(3-methylphenyl)methoxy]benzoic acid

  • MDL number MFCD08444968
  • Cas Number : 923132-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27389
Availability details for
Quantity
  • Availability
  • Add to Cart