ENA116261638 Enamine

5-(chloromethyl)-3-isopropyl-1,2,4-oxadiazole

  • MDL number MFCD08691639
  • Cas Number : 54042-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Country of Origin Ukraine

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27390
Availability details for
Quantity
  • Availability
  • Add to Cart