ENA116261639 Enamine

5-(chloromethyl)-3-ethyl-1,2,4-oxadiazole

  • MDL number MFCD08059891
  • Cas Number : 50737-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27391

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27391
Availability details for
Quantity
  • Availability
  • Add to Cart