ENA116261643 Enamine

1-(chloromethyl)-2-(difluoromethoxy)benzene

  • MDL number MFCD08691303
  • Cas Number : 42145-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26360
Availability details for
Quantity
  • Availability
  • Add to Cart