ENA116261665 Enamine

3-({[2-(4-methylphenyl)vinyl]sulfonyl}amino)propanoic acid

  • MDL number MFCD06671218
  • Cas Number : 851115-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13284

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13284
Availability details for
Quantity
  • Availability
  • Add to Cart