ENA116261666 Enamine

(4-nitrophenyl)methanesulfonyl chloride

  • MDL number MFCD00034030
  • Cas Number : 4025-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88...90
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27238

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-27238
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart