ENA116261672 Enamine

5-[(2-oxo-1,2-dihydropyridin-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD08444520
  • Cas Number : 923216-10-8
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27242
Availability details for
Quantity
  • Availability
  • Add to Cart