ENA116261691 Enamine

N-methyl-1,3-benzoxazol-2-amine

  • MDL number MFCD00613415
  • Cas Number : 19776-98-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27250

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27250
Availability details for
Quantity
  • Availability
  • Add to Cart