ENA116261703 Enamine

4-(6-methyl-1,3-benzothiazol-2-yl)aniline

  • MDL number MFCD00005780
  • Cas Number : 92-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194...196
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17374

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-17374
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart