ENA116261735 Enamine

3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoic acid

  • MDL number MFCD00017519
  • Cas Number : 20170-32-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165...167
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-17562

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-17562
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart