ENA116261744 Enamine

5-(chloromethyl)-3-(2-methylpropyl)-1,2,4-oxadiazole

  • MDL number MFCD09040567
  • Cas Number : 929973-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27272
Availability details for
Quantity
  • Availability
  • Add to Cart