ENA116261759 Enamine

2-[(pyridin-2-yl)methoxy]benzaldehyde

  • MDL number MFCD03110642
  • Cas Number : 54402-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-71
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27286

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27286
Availability details for
Quantity
  • Availability
  • Add to Cart