ENA116261766 Enamine

2-[(2,2,2-trifluoroethyl)thio]nicotinic acid

  • MDL number MFCD09040575
  • Cas Number : 929973-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27294

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27294
Availability details for
Quantity
  • Availability
  • Add to Cart