ENA116261771 Enamine

5H,10H-imidazo[1,2-b]isoquinoline-5,10-dione

  • MDL number MFCD00128522
  • Cas Number : 36142-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27296
Availability details for
Quantity
  • Availability
  • Add to Cart