ENA116261799 Enamine

3-(2H-1,2,3-triazol-2-yl)propanenitrile

  • MDL number MFCD09040577
  • Cas Number : 4320-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27315
Availability details for
Quantity
  • Availability
  • Add to Cart