ENA116261806 Enamine

3-butyl-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD09040580
  • Cas Number : 884058-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27326
Availability details for
Quantity
  • Availability
  • Add to Cart