ENA116261808 Enamine

2-chloro-N-{[(3,4-difluorophenyl)carbamoyl]methyl}acetamide

  • MDL number MFCD09040353
  • Cas Number : 929973-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23132

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23132
Availability details for
Quantity
  • Availability
  • Add to Cart