ENA116261821 Enamine

3-amino-N-cyclopropyl-4-methoxybenzene-1-sulfonamide

  • MDL number MFCD09040586
  • Cas Number : 929973-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27344
Availability details for
Quantity
  • Availability
  • Add to Cart