ENA116261825 Enamine

2-(furan-2-yl)-4,5-dimethylthieno[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD09040588
  • Cas Number : 929973-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27349
Availability details for
Quantity
  • Availability
  • Add to Cart