ENA116261826 Enamine

3-[2-chloro-N-(2-methoxyphenyl)acetamido]propanamide

  • MDL number MFCD09040372
  • Cas Number : 929973-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-23725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23725
Availability details for
Quantity
  • Availability
  • Add to Cart