ENA116261831 Enamine

3-(1H-1,2,3-triazol-1-yl)propanenitrile

  • MDL number MFCD09040593
  • Cas Number : 4320-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27354
Availability details for
Quantity
  • Availability
  • Add to Cart