ENA116261833 Enamine

2-{[7-(methoxycarbonyl)-4-oxo-3,4-dihydroquinazolin-2-yl]methyl}benzoic acid

  • MDL number MFCD02708127
  • Cas Number : 327091-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27355

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27355
Availability details for
Quantity
  • Availability
  • Add to Cart