ENA116261836 Enamine

N-(tetrahydrofuran-2-ylmethyl)-4-[3-(trifluoromethyl)phenyl]-1,3-thiazol-2-amine

  • MDL number MFCD09040595
  • Cas Number : 929973-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27360

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27360
Availability details for
Quantity
  • Availability
  • Add to Cart