ENA116261844 Enamine

2-{[2-(cyclopropylamino)-2-oxoethyl]thio}benzoic acid

  • MDL number MFCD06660856
  • Cas Number : 926258-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27367

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27367
Availability details for
Quantity
  • Availability
  • Add to Cart