ENA116261845 Enamine

2-methyl-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidin-5-one

  • MDL number MFCD02703191
  • Cas Number : 380391-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27368
Availability details for
Quantity
  • Availability
  • Add to Cart