ENA116261849 Enamine

methyl 2-oxo-1,1',2,2',5',6',7',7'a-octahydrospiro[indole-3,3'-pyrrolizine]-2'-carboxylate

  • MDL number MFCD09040598
  • Cas Number : 1265964-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27373
Availability details for
Quantity
  • Availability
  • Add to Cart