ENA116261850 Enamine

4-hydroxy-3-nitrobenzene-1-sulfonamide

  • MDL number MFCD00129906
  • Cas Number : 24855-58-1
  • Molecular Weight : 218.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27374
Availability details for
Quantity
  • Availability
  • Add to Cart