ENA117737819 Enamine

2-chloro-N-{[4-(dimethylamino)phenyl]methyl}-N-[(oxolan-2-yl)methyl]acetamide

  • MDL number MFCD09702167
  • Cas Number : 1000930-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22877
Availability details for
Quantity
  • Availability
  • Add to Cart