ENA117737820 Enamine

3-[(thien-3-ylcarbonyl)amino]propanoic acid

  • MDL number MFCD09388775
  • Cas Number : 940781-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22997

Safety Information

msds https://www.enaminestore.com/catalog/EN300-22997
Availability details for
Quantity
  • Availability
  • Add to Cart