ENA117737829 Enamine

2-chloro-N-[1-(4-methoxyphenyl)-2-methylpropyl]acetamide

  • MDL number MFCD09403605
  • Cas Number : 941165-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24587
Availability details for
Quantity
  • Availability
  • Add to Cart