ENA117737833 Enamine

2-chloro-N-[1-(4-fluorophenyl)ethyl]-N-methylacetamide

  • MDL number MFCD09403618
  • Cas Number : 941226-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24643
Availability details for
Quantity
  • Availability
  • Add to Cart