ENA117737836 Enamine

ethyl 5-(2-chloroacetamido)-3-methylthiophene-2-carboxylate

  • MDL number MFCD09396667
  • Cas Number : 285138-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24664
Availability details for
Quantity
  • Availability
  • Add to Cart