ENA117737845 Enamine

6-chloro-2-hydroxy-3-methyl-4H-chromen-4-one

  • MDL number MFCD26143734
  • Cas Number : 1934394-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27713
Availability details for
Quantity
  • Availability
  • Add to Cart