ENA117737846 Enamine

5-methyl-2-(piperazin-1-yl)-7H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-7-one

  • MDL number MFCD09388787
  • Cas Number : 940242-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27723

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27723
Availability details for
Quantity
  • Availability
  • Add to Cart