ENA117737849 Enamine

1-[2-fluoro-6-(1H-imidazol-1-yl)phenyl]ethan-1-one

  • MDL number MFCD09388774
  • Cas Number : 940841-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27729
Availability details for
Quantity
  • Availability
  • Add to Cart